Merck
Effects of the herbicide chlorthiamid on the olfactory mucosa.
Toxicology letters (1995-04-01)
C Eriksson, E B Brittebo
ABSTRACT
Chlorthiamid (2,6-dichlorothiobenzamide) and its major metabolite 2,6-dichlorobenzonitrile are olfactory toxicants with a high in vivo covalent binding in the olfactory mucosa of mice. This study showed that the cytochrome P450 (P450) inhibitors, metyrapone and sodium-diethyldithiocarbamate, abolished the chlorthiamid-induced toxicity (12 mg/kg; 0.06 mmol/kg) in C57B1/6 mice suggesting a P450-dependent toxicity. Incubation of [14C]-labelled chlorthiamid with rat olfactory microsomes showed a low NADPH-dependent oxidative covalent binding which was only 3-fold higher than that in liver microsomes. Thus the results do not support a major in situ metabolic activation of chlorthiamid and it is suggested that metabolic activation of the major chlorthiamid metabolite (2,6-dichlorobenzonitrile) is responsible for most of the covalent binding and toxicity of chlorthiamid at this site in vivo. Thiobenzamide (16 mg/kg; 0.12 mmol/kg), a dechlorinated chlorthiamid-analog, induced no marked morphological changes in the olfactory mucosa demonstrating that chlorines in the 2,6-position are important for the chlorthiamid-induced toxicity at this site.
Product Description
Supelco
Chlorthiamid, PESTANAL®, analytical standard
Sigma-Aldrich
Thiobenzamide, 98%