Merck
Metabolism and covalent binding to DNA of 7-methylbenzo(a)pyrene.
Cancer research (1982-10-01)
T Kinoshita, M Konieczny, R Santella, A M Jeffrey
ABSTRACT
The ultimate carcinogenic form of benzo(a)pyrene (BP) is thought to result from metabolic activation at the 7 to 10 positions. Substitution by a methyl group at these positions would be expected to inhibit strongly their metabolism even though 7-methylbenzo(a)pyrene (7-MeBP) has been reported to be carcinogenic in some tumor models. The metabolism of 7-MeBP was, therefore, studied using both microsomal preparations and whole cells, the products being analyzed by high-pressure liquid chromatography, fluorescence spectrophotometry, and mass spectrometry. These studies revealed that many of the expected metabolites were formed by microsomes, but in addition 7-MeBP yielded a compound which was isolated and identified as trans-7,8-dihydro-7,8-dihydroxy-7-methylbenzo(a)pyrene. These results indicate that, despite the presence of a methyl group at the 7 position, a substituted BP can undergo the same initial metabolic activation as BP itself. However, in contrast to BP, the 7,8-dihydrodiol formed from 7-MeBP was almost racemic, and neither enantiomer was very active in the Ames bacterial mutagenesis assay when compared with trans-7,8-dihydro-7,8-dihydroxybenzo(a)pyrene. The metabolism of 7-MeBP was also studied in 10T1/2 cells. The hydrocarbon was metabolized readily and bound to DNA of the cells to about one-eighth of the level found for BP. However, no 7,8-dihydro-7,8-dihydroxy-7-methylbenzo(a)pyrene could be detected in the culture medium.
Sigma-Aldrich
7-Methylbenzo[a]pyrene, 98%