Merck
  • Disposition of the hypolipidaemic agent, 2,3-dihydrophthalazine-1,4-dione, in Sprague Dawley rats.
Disposition of the hypolipidaemic agent, 2,3-dihydrophthalazine-1,4-dione, in Sprague Dawley rats.
The Journal of pharmacy and pharmacology (1989-06-01)
I H Hall, C B Oswald, S D Wyrick, J H Maguire, R P Shrewsbury
ABSTRACT
The disposition of [14C]2,3-dihydrophthalazine-1,4-dione, a potent hypolipidaemic agent, has been determined after both intravenous and oral administration. Both the routes of administration afforded multi-exponential disposition with an estimated t1/2 of approximately 75 h. After oral administration, the drug was observed to be absorbed rapidly from the intestine and distributed quickly to all tissues of the body. A large quantity of the 14C-radioactivity was found in the skin and carcass. Approximately 35% of the administered radioactivity was excreted in urine after oral administration and 11% in the faeces. Approximately 66% of the radioactivity excreted in urine was the parent drug. There was evidence of an additional metabolite which accounted for 28% of the urinary radioactive excretion. The parent drug has little serum protein binding, is highly water soluble, and is probably taken up by cells by passive diffusion.
Sigma-Aldrich
Phthalhydrazide, ReagentPlus®, 99%