Merck
  • Effects of pyrimidine nucleoside phosphorylase inhibitors on hepatic fluoropyrimidine elimination in the rat.
Effects of pyrimidine nucleoside phosphorylase inhibitors on hepatic fluoropyrimidine elimination in the rat.
Cancer research (1989-05-15)
F P LaCreta, B S Warren, W M Williams
ABSTRACT
The breakdown of 5-fluoro-2'-deoxyuridine (FdUrd) to 5-fluorouracil (FUra) is catalyzed by the pyrimidine nucleoside phosphorylases, uridine phosphorylase and thymidine phosphorylase. The effects of nucleoside phosphorylase inhibitors on FdUrd and FUra elimination by the isolated perfused rat liver were investigated. The inhibitor was injected into the perfusion reservoir 5 min before FdUrd or FUra, and serial perfusion fluid samples were collected for fluoropyrimidine analysis. The disappearance of each fluoropyrimidine followed Michaelis-Menten kinetics, as shown previously. 6-Benzyl-2-thiouracil, a thymidine phosphorylase-selective inhibitor, and 1-(2'-deoxy-beta-D-glucopyranosyl)thymine, a uridine phosphorylase-selective inhibitor, each decreased the rate of FdUrd disappearance (apparent Ki, 1.4-1.6 and 3.8 mM, respectively) but had no direct effect on FUra disappearance. However, 6-benzyl-2-thiouracil decreased the peak concentration of FUra derived from administered FdUrd and increased the t 1/2 of disappearance of derived FUra due to its delayed formation. 2,6-Dihydroxypyridine, a uridine phosphorylase-selective inhibitor, decreased the rate of FdUrd disappearance (apparent Ki, 12.4-16.2 microM) and directly inhibited FUra elimination (apparent Ki, 4.3-5.3 microM). 2,4-Dihydroxypyridine, which does not inhibit pyrimidine nucleoside phosphorylases, directly inhibited FUra elimination (apparent Ki, 77 microM) and also decreased the rate of FdUrd disappearance, possibly due to product (FUra) inhibition. It was concluded that the hepatic elimination of FdUrd is slowed by pyrimidine nucleoside phosphorylase inhibitors and that some of these drugs block FUra, as well as FdUrd, elimination.
Sigma-Aldrich
2,4-Dihydroxypyridine, 97%