Merck
Magnetic resonance frequency shifts during acute MS lesion formation.
Neurology (2013-06-14)
Vanessa Wiggermann, Enedino Hernández Torres, Irene M Vavasour, G R Wayne Moore, Cornelia Laule, Alex L MacKay, David K B Li, Anthony Traboulsee, Alexander Rauscher
ABSTRACT
We investigated the evolution of new multiple sclerosis (MS) lesions over time using frequency shifts of the magnetic resonance (MR) signal. Twenty patients with relapsing-remitting MS were serially scanned for 6 months at 1-month intervals. Maps of MR frequency shifts were acquired using susceptibility-weighted imaging. New lesions were identified by enhancement with gadolinium (Gd). Forty new lesions were identified as areas of signal increase on Gd-enhanced scans. Up to 3 months before lesion appearance, the frequency in areas of future Gd enhancement was not detectably different from the frequency in normal-appearing white matter. Rapid increase in MR frequency was observed between 1 month before and 1 month after Gd enhancement. Two months postenhancement and later, the frequency stabilized and remained at a constantly increased level. These findings suggest that an increase in MR frequency does not simply reflect blood-brain barrier disruption or edema; rather, it reflects a change of tissue architecture as a consequence of new lesion formation. The data demonstrate that the MR frequency of focal MS lesions is increased before the lesions appear on conventional MRI. Unlike many other advanced imaging techniques, the images for frequency mapping can be rapidly acquired at high spatial resolution and standardized on most clinical scanners.
Sigma-Aldrich
Gadolinium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Gadolinium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Sigma-Aldrich
Gadolinium, chips
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis