Merck
  • Improvement and formation of UV-induced damage on LBO crystal surface during long-term high-power third-harmonic generation.
Improvement and formation of UV-induced damage on LBO crystal surface during long-term high-power third-harmonic generation.
Optics express (2013-04-03)
Hailong Hong, Qiang Liu, Lei Huang, Mali Gong
ABSTRACT
We demonstrate the improvement and formation of UV-induced damage on LBO crystal output surface during long-term (130 h) high-power (20 W) high-repetition-rate (80 kHz) third-harmonic generation. The output surface was super-polished (RMS surface roughness <0.6 nm) to sub-nanometer scale super smooth roughness. The surface lifetime has been improved more than 20-fold compared with the as-polished ones (RMS surface roughness 4.0~8.0 nm). The damage could be attributed to the consequence of thermal effects resulted from impurity absorptions. Simultaneously, it was verified that the impurities originated in part from the UV-induced deposition.
Sigma-Aldrich
Lithium tetraborate, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium tetraborate, ≥99.995% trace metals basis