Merck
  • Multicolor stimulated Raman scattering microscopy with a rapidly tunable optical parametric oscillator.
Multicolor stimulated Raman scattering microscopy with a rapidly tunable optical parametric oscillator.
Optics letters (2013-03-05)
Lingjie Kong, Minbiao Ji, Gary R Holtom, Dan Fu, Christian W Freudiger, X Sunney Xie
ABSTRACT
Stimulated Raman scattering (SRS) microscopy allows label-free chemical imaging based on vibrational spectroscopy. Narrowband excitation with picosecond lasers creates the highest signal levels and enables imaging speeds up to video-rate, but it sacrifices chemical specificity in samples with overlapping bands compared to broadband (multiplex) excitation. We develop a rapidly tunable picosecond optical parametric oscillator with an electro-optical tunable Lyot filter, and demonstrate multicolor SRS microscopy with synchronized line-by-line wavelength tuning to avoid spectral artifacts due to sample movement. We show sensitive imaging of three different kinds of polymer beads and live HeLa cells with moving intracellular lipid droplets.
Product Description
Supelco
Melamine, analytical standard
Sigma-Aldrich
Melamine, 99%
Supelco
Melamine, pharmaceutical impurity standard, ≥95.0% (HPLC)
Supelco
Melamine, Pharmaceutical Secondary Standard; Certified Reference Material