Merck
  • Nd(2)Fe(14)B/FeCo anisotropic nanocomposite films with a large maximum energy product.
Nd(2)Fe(14)B/FeCo anisotropic nanocomposite films with a large maximum energy product.
Advanced materials (Deerfield Beach, Fla.) (2012-10-31)
Wei-Bin Cui, Yukiko K Takahashi, Kazuhiro Hono
ABSTRACT
Anisotropic Nd(2) Fe(14) B/Fe(67) Co(33) nanocomposite thin films are successfully fabricated. The multilayer composite films comprise Nd-rich shell-enveloped Nd(2) Fe(14) B grains and a Fe(67) Co(33) phase. The strong (001) texture of the Nd(2) Fe(14) B grains and the presence of exchange-coupled Fe(67) Co(33) lead to a high remanence and the presence of the Nd-rich shell gives rise to a high coercivity. The unique nanocomposite microstructure provides hints for developing rare-earth-lean high-performance magnets.
Sigma-Aldrich
Neodymium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis
Sigma-Aldrich
Neodymium, ingot (in mineral oil), 99.9% trace rare earth metals basis