Merck
  • Antibacterial activities of Nd doped and Ag coated TiO2 nanoparticles under solar light irradiation.
Antibacterial activities of Nd doped and Ag coated TiO2 nanoparticles under solar light irradiation.
Colloids and surfaces. B, Biointerfaces (2012-09-27)
Anuja Bokare, Avinash Sanap, Mrinal Pai, Sushma Sabharwal, Anjali A Athawale
ABSTRACT
Nanosized (8-9 nm) Nd doped and Ag coated TiO(2) nanoparticles have been synthesized by sol-gel method. The physicochemical properties of these particles were investigated by X-ray diffraction (XRD), diffuse reflectance UV-visible (DRUV) spectra and Brunauer-Emmett-Teller (BET) surface area analysis. The antibacterial activities of the samples were studied for Staphylococcus aureus (S. aureus) and Escherichia coli (E. coli) both, under the light and dark conditions. The results reveal that the extent of activity shows the order as undoped TiO(2)<Nd doped TiO(2)<Ag coated TiO(2). The mechanism of bactericidal action of the nanoparticles, in presence of sunlight has been explained with the help of microscopic analyses. The bacterial damage is observed to proceed through initial perforation of the cell, damage of cell wall and finally the bacterial death.
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis
Sigma-Aldrich
Neodymium, ingot (in mineral oil), 99.9% trace rare earth metals basis
Sigma-Aldrich
Neodymium, chips, 99.9% trace rare earth metals basis