Merck
  • Behavior of almond oil bodies during in vitro gastric and intestinal digestion.
Behavior of almond oil bodies during in vitro gastric and intestinal digestion.
Food & function (2012-02-23)
Sophie Gallier, Harjinder Singh
ABSTRACT
An aqueous suspension of almond oil bodies (about 10% lipids) was prepared and subjected to in vitro gastric (with pepsin) and intestinal (with bile salts and pancreatin) digestion, simulating fasting conditions. The physicochemical and structural changes of the almond oil body emulsion were examined. The almond oil body emulsion behaved similarly to a protein-stabilized emulsion, with flocculation of the oil bodies occurring under gastric conditions. Proteins, peptides, and phospholipids covered the surface of the oil bodies throughout gastric digestion. Under intestinal conditions, bile salts displaced the interfacial peptides and phospholipids, and disrupted the flocs. Gastric pepsinolysis of almond proteins was a prerequisite for their digestion in the duodenum. The oil body membrane had a negative impact on the efficiency of gastric digestion, and long chain fatty acids, the main lipolytic products, accumulated at the surface of the oil bodies and therefore limited the activity of pancreatic lipase.
Sigma-Aldrich
Almond oil, sweet, Certified organic (NOP)
Sigma-Aldrich
Almond oil from Prunus dulcis, tested according to Ph. Eur.