Merck
  • Formation and structure elucidation of N-(2,3,4-tri-O-acetyl-beta-D-glucopyranosyl)-N'-acetylthiourea.
Formation and structure elucidation of N-(2,3,4-tri-O-acetyl-beta-D-glucopyranosyl)-N'-acetylthiourea.
Carbohydrate research (2008-11-04)
László Somogyi, Gyula Batta
ABSTRACT
Treatment with concd HCl/MeOH transformed N-(tetra-O-acetyl-beta-D-glucopyranosyl)-N'-acetylthiourea, via selective cleavage of the primary alcoholic ester group, into the title compound.
Sigma-Aldrich
Acetylthiourea, 99%