Merck
  • Assessing intermittent pesticide exposure from flea control collars containing the organophosphorus insecticide tetrachlorvinphos.
Assessing intermittent pesticide exposure from flea control collars containing the organophosphorus insecticide tetrachlorvinphos.
Journal of exposure science & environmental epidemiology (2008-01-03)
M Keith Davis, J Scott Boone, John E Moran, John W Tyler, Janice E Chambers
ABSTRACT
Fleas are a persistent problem for pets that require implementation of control measures. Consequently, pesticide use by homeowners for flea control is common and may increase pesticide exposure for adults and children. Fifty-five pet dogs (23 in study 1; 22 in study 2) of different breeds and weights were treated with over-the-counter flea collars containing tetrachlorvinphos (TCVP). During study 1, fur of treated dogs was monitored for transferable TCVP residues using cotton gloves to pet the dogs during 5-min rubbings post-collar application. Plasma cholinesterase (ChE) activity was also measured in treated dogs. Average amounts of TCVP transferred from the fur of the neck (rubbing over the collar) and from the back to gloves at 3 days post-collar application were 23,700+/-2100 and 260+/-50 microg/glove, respectively. No inhibition of plasma ChE was observed. During study 2, transferable TCVP residues to cotton gloves were monitored during 5-min rubbings post-collar application. Transferable residues were also monitored on cotton tee shirts worn by children and in the first morning urine samples obtained from adults and children. Average amounts of TCVP transferred to gloves at 5 days post-collar application from the neck (over the collar) and from the back were 22,400+/-2900 and 80+/-20 microg/glove, respectively. Tee shirts worn by children on days 7-11 contained 1.8+/-0.8 microg TCVP/g shirt. No significant differences were observed between adults and children in urinary 2,4,5-trichloromandelic acid (TCMA) levels; however, all TCMA residues (adults and children) were significantly greater than pretreatment concentrations (alpha=0.05). The lack of ChE inhibition in dogs and the low acute toxicity level of TCVP (rat oral LD(50) of 4-5 g/kg) strongly suggest that TCVP is rapidly detoxified and excreted and therefore poses a very low toxicological risk, despite these high residues.
Sigma-Aldrich
2,3,6-Trichlorobenzaldehyde, 97%
Pricing and availability is not currently available.