- Endothelin-binding inhibitors, BE-18257A and BE-18257B. I. Taxonomy, fermentation, isolation and characterization.
Endothelin-binding inhibitors, BE-18257A and BE-18257B. I. Taxonomy, fermentation, isolation and characterization.
The Journal of antibiotics (1991-12-01)
K Kojiri, M Ihara, S Nakajima, K Kawamura, K Funaishi, M Yano, H Suda
PMID1778786
ABSTRACT
Two endothelin (ET)-binding inhibitors, BE-18257A and BE-18257B, which antagonized 125I-ET-1 binding to a porcine aortic smooth muscle membrane, were isolated from the mycelium of a strain of Streptomyces misakiensis. These binding inhibitors were extracted with methanol from mycelium and purified by HPLC.