- Suicide by multidrug ingestion: hypothesis on the role played by the self-administration of activated charcoal.
Suicide by multidrug ingestion: hypothesis on the role played by the self-administration of activated charcoal.
The American journal of forensic medicine and pathology (2007-02-28)
D R Vantaggiato, N De Giovanni
PMID17325466
ABSTRACT
A fatal suicidal ingestion of drugs, together with activated charcoal, is reported. The death occurred 31 hours after the self-administration. The autopsy revealed a large amount of gastric content that appeared to be a compact mass of black color. Toxicologic analyses showed the presence of toxic levels of desalkylflurazepam and trazodone; metamizole and pridinol were also detected. The obtained results supported the hypothesis of a death due to acute intoxication delayed by the self-administration of activated charcoal, which elimination was probably hindered by the action of pridinol.