Merck
  • Organic nanocrystals with tunable morphologies and optical properties prepared through a sonication technique.
Organic nanocrystals with tunable morphologies and optical properties prepared through a sonication technique.
Physical chemistry chemical physics : PCCP (2006-07-13)
Yong Sheng Zhao, Wensheng Yang, Jiannian Yao
ABSTRACT
Nanocrystals with tunable morphologies and optical properties were successfully fabricated from an organic functional low-molecular-weight compound, 2,4,5-triphenylimidazole (lophine), through a sonication technique.
Sigma-Aldrich
2,4,5-Triphenylimidazole, 98%