Merck
  • Sorting of polymorphs based on mechanical properties. Trimorphs of 6-chloro-2,4-dinitroaniline.
Sorting of polymorphs based on mechanical properties. Trimorphs of 6-chloro-2,4-dinitroaniline.
Chemical communications (Cambridge, England) (2005-05-12)
C Malla Reddy, Srinivas Basavoju, Gautam R Desiraju
ABSTRACT
Mechanical shearing of crystals of a polymorph with a layered structure may be used to separate them from harder crystals of a visually indistinguishable polymorph that resist such shearing.
Sigma-Aldrich
6-Chloro-2,4-dinitroaniline, 97%