Merck
  • Effect of dietary high-oleic-acid oils that are rich in antioxidants on microsomal lipid peroxidation in rats.
Effect of dietary high-oleic-acid oils that are rich in antioxidants on microsomal lipid peroxidation in rats.
Journal of agricultural and food chemistry (2005-02-03)
Javier S Perona, Cesar Arcemis, Valentina Ruiz-Gutierrez, Angel Catalá
ABSTRACT
In contrast to other metabolic functions, the role of dietary antioxidants and oil on microsomal lipid oxidation has been less extensively studied. This study examines ascorbate-Fe(2+) and NADPH-induced lipid peroxidation of hepatic microsomes of rats that were fed for three weeks high-oleic-acid oils (high-oleic sunflower oil, HOSO; olive oil, OO; or olive pomace oil, OPO) containing different concentrations of the antioxidants alpha-tocopherol, erythrodiol, and oleanolic acid. The fatty acid composition of hepatic microsomes of Wistar rats that were fed for three weeks with the above-mentioned oils had lower proportions of C16:0, C18:2n6, and C22:6n3 and higher proportions of C18:0 and C18:1n9 than rats fed the control diet. Light emission by hepatic microsomes increased, in the first 180 min, 2-fold after ascorbate-Fe(2+) addition compared with NADPH addition. Both increases were less pronounced in microsomes of OPO-fed rats and to a smaller extent in microsomes of OO-fed rats. Smaller increases in light emission were associated positively with higher concentrations of dietary alpha-tocopherol, erythrodiol, and oleanolic acid but were not associated with changes in fatty acid composition of hepatic microsomes. Addition of alpha-tocopherol, erythrodiol, or oleanolic acid decreased light emission of hepatic microsomes with a greater inhibition in microsomes of rats fed the control diet. Our data suggest that erythrodiol and oleanolic acids partly explain the protective effect of dietary OPO on microsomal lipid peroxidation in rats.