Merck
  • Evidence for the combined participation of a C10 and a C15 precursor in the biosynthesis of moenocinol, the lipid part of the moenomycin antibiotics.
Evidence for the combined participation of a C10 and a C15 precursor in the biosynthesis of moenocinol, the lipid part of the moenomycin antibiotics.
Chembiochem : a European journal of chemical biology (2003-11-13)
Ines Neundorf, Christian Köhler, Lothar Hennig, Matthias Findeisen, Duilio Arigoni, Peter Welzel
ABSTRACT
Upon feeding of [2-(13)C,4-(2)H]-1-deoxy-D-xylulose to Streptomyces ghanaensis, the deuterium label was retained exclusively at positions C-7 and C-17 in the moenocinol part of the moenomycin antibiotics. This result vindicates the hypothesis that the C(25) structure of moenocinol is assembled from a C(10) and a C(15) precursor, each of which requires for its formation the involvement of a dimethylallyl diphosphate starter unit.
Sigma-Aldrich
1-Deoxy-D-xylulose, ≥80% (TLC)