Merck
  • Reducing-equivalent transfer to the mitochondria during gluconeogenesis and ureogenesis in hepatocytes from rats of different thyroid status.
Reducing-equivalent transfer to the mitochondria during gluconeogenesis and ureogenesis in hepatocytes from rats of different thyroid status.
Biochimica et biophysica acta (1992-10-06)
R B Gregory, J W Phillips, M N Berry
ABSTRACT
Isolated hepatocytes from hypothyroid, euthyroid and hyperthyroid rats have been employed to investigate the relative importance of reducing-equivalent shuttles for the transfer of hydrogen between cytoplasm and mitochondria during simultaneous ureogenesis and gluconeogenesis. In cells from hypothyroid animals, a 58% depression of glucose formation and 68% reduction in ureogenesis were induced by n-butylmalonate, an inhibitor of the malate shuttle. A more reduced state of the cytoplasmic compartment and a substantial fall in the concentrations of pyruvate, aspartate, alanine and glutamate accompanied this inhibition. Preincubation of cells with n-butylmalonate yielded greater inhibitory effects than observed in the absence of preincubation. The inhibitory effects on gluconeogenesis and ureogenesis were less in cells from euthyroid rats and were very much reduced in the case of glucose synthesis and absent in the case of ureogenesis, in cells from hyperthyroid rats. It is inferred that both the malate-aspartate and alpha-glycerophosphate shuttles may function in the transfer of reducing equivalents from cytoplasm to mitochondria during ureogenesis in hepatocytes. The major inhibition by n-butylmalonate of glucose and urea synthesis in hepatocytes from hypothyroid rats is due to the diminished activity of the alpha-glycerophosphate shuttle in these cells. Moreover, it follows that the NADH arising from the cytoplasmic malate dehydrogenase-catalysed reaction is accessible to both the malate-aspartate shuttle and the alpha-glycerophosphate shuttle.
Sigma-Aldrich
Butylmalonic acid, 99%