Merck
  • Poly(ADP-ribose) polymerase (PARP-1) has a controlling role in homologous recombination.
Poly(ADP-ribose) polymerase (PARP-1) has a controlling role in homologous recombination.
Nucleic acids research (2003-08-22)
Niklas Schultz, Elena Lopez, Nasrollah Saleh-Gohari, Thomas Helleday
ABSTRACT
Cells with non-functional poly(ADP-ribose) polymerase (PARP-1) show increased levels of sister chromatid exchange, suggesting a hyper recombination phenotype in these cells. To further investigate the involvement of PARP-1 in homologous recombination (HR) we investigated how PARP-1 affects nuclear HR sites (Rad51 foci) and HR repair of an endonuclease-induced DNA double-strand break (DSB). Several proteins involved in HR localise to Rad51 foci and HR-deficient cells fail to form Rad51 foci in response to DNA damage. Here, we show that PARP-1 mainly does not localise to Rad51 foci and that Rad51 foci form in PARP-1-/- cells, also in response to hydroxyurea. Furthermore, we show that homology directed repair following induction of a site-specific DSB is normal in PARP-1-inhibited cells. In contrast, inhibition or loss of PARP-1 increases spontaneous Rad51 foci formation, confirming a hyper recombination phenotype in these cells. Our data suggest that PARP-1 controls DNA damage recognised by HR and that it is not involved in executing HR as such.
Sigma-Aldrich
1,5-Isoquinolinediol, ≥98% (HPLC), powder