- Pyrrolizidine alkaloids from Senecio lorenthii.
Pyrrolizidine alkaloids from Senecio lorenthii.
Fitoterapia (2001-07-14)
M Noorwala, F V Mohammad, V U Ahmad, B Sener, F Ergun, D Deliorman
PMID11449528
ABSTRACT
The isolation and NMR spectra of otosenine (1) and seneciphylline (2) from Senecio lorenthii are reported.
MATERIALS