Merck
  • Enantioselective supercritical fluid chromatography using ristocetin A chiral stationary phases.
Enantioselective supercritical fluid chromatography using ristocetin A chiral stationary phases.
The Analyst (2000-08-10)
L A Svensson, P K Owens
ABSTRACT
Racemic mixtures of five acidic drugs have been successfully separated by supercritical fluid chromatography (SFC) using macrocyclic antibiotic chiral stationary phases (CSPs). A ristocetin A CSP has been prepared 'in-house' and effectively applied in packed capillary SFC to separate the enantiomers of dichlorprop (R(s) = 1.4), ketoprofen (R(s) = 0.9) and warfarin (R(s) = 0.9). The commercial ristocetin A CSP (Chirobiotic R) was subsequently studied in packed column SFC with similar results where the enantiomers of warfarin (R(s) = 2.2), coumachlor (R(s) = 2.5) and thalidomide (R(s) = 0.6) were separated. Interestingly, differences were observed between the two differently immobilised CSPs where the enantiomers of dichlorprop and ketoprofen, which were separated on the 'in-house' CSP, could not be separated on the commercial phase.
Sigma-Aldrich
Coumachlor, 98%