MilliporeSigma
  • Molecular moment similarity between several nucleoside analogs of thymidine and thymidine. sil@watson.ibm.com.
Molecular moment similarity between several nucleoside analogs of thymidine and thymidine. sil@watson.ibm.com.
Journal of biomolecular structure & dynamics (1999-08-14)
B D Silverman, M C Pitman, D E Platt
ABSTRACT
Molecular moment descriptors of the shape and charge distributions of twenty five nucleoside structures have been examined. The structures include thymidine as well as the difluorotoluene nucleoside analog which has been found to pair efficiently with adenine by polymerase catalysis. The remaining twenty three structures have been chosen to be as structurally similar to thymidine and to the difluorotoluene nucleoside analog as possible. The moment descriptors which include a description of the relationship of molecular charge to shape show the difluorotoluene nucleoside to be one of the most proximate molecules to thymidine in the space of the molecular moments. The calculations, therefore, suggest that polymerase specificity might be not only a consequence of molecular steric features alone but also of the molecular electrostatic environment and its registration with molecular shape.
Sigma-Aldrich
2,4-Difluorotoluene, 99%