Merck
  • Coenzyme A-disulfide reductase from Staphylococcus aureus: evidence for asymmetric behavior on interaction with pyridine nucleotides.
Coenzyme A-disulfide reductase from Staphylococcus aureus: evidence for asymmetric behavior on interaction with pyridine nucleotides.
Biochemistry (1999-03-03)
J Luba, V Charrier, A Claiborne
ABSTRACT
An unusual flavoprotein disulfide reductase, which catalyzes the NADPH-dependent reduction of CoASSCoA, has recently been purified from the human pathogen Staphylococcus aureus [delCardayré, S. B., Stock, K. P., Newton, G. L., Fahey, R. C., and Davies, J. E. (1998) J. Biol. Chem. 273, 5744-5751]. Coenzyme A-disulfide reductase (CoADR) lacks the redox-active protein disulfide characteristic of the disulfide reductases; instead, NADPH reduction yields 1 protein-SH and 1 CoASH. Furthermore, the CoADR sequence reveals the presence of a single putative active-site Cys (Cys43) within an SFXXC motif also seen in the Enterococcus faecalis NADH oxidase and NADH peroxidase, which use a single redox-active cysteine-sulfenic acid in catalysis. In this report, we provide a detailed examination of the equilibrium properties of both wild-type and C43S CoADRs, focusing on the role of Cys43 in the catalytic redox cycle, the behavior of both enzyme forms on reduction with dithionite and NADPH, and the interaction of NADP+ with the corresponding reduced enzyme species. The results of these analyses, combined with electrospray mass spectrometric data for the two oxidized enzyme forms, fully support the catalytic redox role proposed for Cys43 and confirm that this is the attachment site for bound CoASH. In addition, we provide evidence indicating dramatic thermodynamic inequivalence between the two active sites per dimer, similar to that documented for the related enzymes mercuric reductase and NADH oxidase; only 1 FAD is reduced with NADPH in wild-type CoADR. The EH2.NADPH/EH4.NADP+ complex which results is reoxidized quantitatively in titrations with CoASSCoA, supporting a possible role for the asymmetric reduced dimer in catalysis.
Sigma-Aldrich
Coenzyme A, oxidized lithium salt, ≥85%
Pricing and availability is not currently available.