Merck
  • Unbiased molecular analysis of T cell receptor expression using template-switch anchored RT-PCR.
Unbiased molecular analysis of T cell receptor expression using template-switch anchored RT-PCR.
Current protocols in immunology (2011-08-03)
Máire F Quigley, Jorge R Almeida, David A Price, Daniel C Douek
ABSTRACT
A detailed knowledge of the principles that guide clonal selection within the memory and effector T cell pools is essential to further our understanding of the factors that influence effective T cell-mediated immunity and has direct implications for the rational design of vaccines and immunotherapies. This unit provides methods for the unbiased quantification and characterization of all expressed T cell receptor (TCR) gene products within any defined T cell population. The approach is based on a template-switch anchored reverse transcription-polymerase chain reaction (RT-PCR) and is optimized for the analysis of antigen-specific T cells isolated directly ex vivo.
Sigma-Aldrich
RNase AWAY®, decontamination reagent for RNase
Pricing and availability is not currently available.