Merck
  • Stereospecific oxidation of (R)- and (S)-1-indanol by naphthalene dioxygenase from Pseudomonas sp. strain NCIB 9816-4.
Stereospecific oxidation of (R)- and (S)-1-indanol by naphthalene dioxygenase from Pseudomonas sp. strain NCIB 9816-4.
Applied and environmental microbiology (1997-05-01)
K Lee, S M Resnick, D T Gibson
ABSTRACT
A recombinant Escherichia coli strain which expresses naphthalene dioxygenase (NDO) from Pseudomonas sp. strain NCIB 9816-4 oxidized (S)-1-indanol to trans-(1S,3S)-indan-1,3-diol (95.5%) and (R)-3-hydroxy-1-indanone (4.5%). The same cells oxidized (R)-1-indanol to cis-1,3-indandiol (71%), (R)-3-hydroxy-1-indanone (18.2%), and cis-1,2,3-indantriol (10.8%). Purified NDO oxidized (S)-1-indenol to both syn- and anti-2,3-dihydroxy-1-indanol.
Sigma-Aldrich
(S)-(+)-1-Indanol, 99%
Sigma-Aldrich
1-Indanol, 98%
Sigma-Aldrich
(R)-(−)-1-Indanol, 99%