Merck
  • Delayed and immediate allergy caused by methylhexahydrophthalic anhydride.
Delayed and immediate allergy caused by methylhexahydrophthalic anhydride.
Contact dermatitis (1997-01-01)
L Kanerva, H Hyry, R Jolanki, M Hytönen, T Estlander
ABSTRACT
Epoxy resin compounds (ERC) include a large number of chemicals, such as epoxy resins (ER), reactive diluents and hardeners. Many hardeners, e.g., aliphatic polyamines, are well-known sensitizers. Another type of ER hardeners are the phthalic anhydrides, such as methylhexahydrophthalic anhydride (MHHPA) and methyltetrahydrophthalic anhydride (MTHPA), which have been reported as causing immunologically-mediated respiratory diseases and contact urticaria, but not allergic contact dermatitis. Here, we present a horizontal boring-machine worker who developed allergic contact dermatitis, as well as allergic rhinitis and an immediate contact skin reaction from MHHPA. Patch testing with a dilution series of MHHPA in pet. elicited the following results: 2%, 1% and 0.5%, +2; 0.25% and 0.125%, + (3- to 6-day readings). An immunohistochemical and electron microscopic study also indicated that the patch test reactions were conventional-delayed allergic reactions. Interleukin 8 was observed in the epidermal cells, whereas interleukin 4 immunoreactivity was detected in the dermal cells. Immunoreactivity to-interleukin 5, granulocyte/macrophage-colophony stimulating factor (GM-CSF) or eosinophil cationic protein was not seen. In conclusion, the patient developed both Type I and Type IV allergy to MHHPA. The clinical data, patch test results, immunohistochemical and electron microscopic observations indicated that the MHHPA allergy detected by the patch test reaction was a conventional delayed-type hypersensitivity reaction. The patient also had an allergic patch test reaction to para-phenylenediamine and diaminodiphenylmethane, possibly representing occupational sensitization.
Sigma-Aldrich
Hexahydro-4-methylphthalic anhydride, mixture of cis and trans, 96%
Pricing and availability is not currently available.