Merck
  • beta-Apo-8'-carotenal, but not beta-carotene, is a strong inducer of liver cytochromes P4501A1 and 1A2 in rat.
beta-Apo-8'-carotenal, but not beta-carotene, is a strong inducer of liver cytochromes P4501A1 and 1A2 in rat.
Xenobiotica; the fate of foreign compounds in biological systems (1996-09-01)
S Gradelet, J Leclerc, M H Siess, P O Astorg
ABSTRACT
1. The catalytic activities of several phase I and II xenobiotic-metabolizing enzymes and their immunochemical detection have been investigated in liver microsomes and cytosol of the male rat, which had been fed for 15 days with diets containing 300 mg/kg beta-carotene isomers (all-trans beta-carotene or beta-carotene from Dunaliella salina rich in 9-cis isomer or isomerized beta-carotene), or apocarotenoids as beta-apo-8'-carotenal, ethyl beta-apo-8'-carotenoate and citranaxanthin. 2. Beta-carotene, either all-trans or containing cis isomers, did not induce any significant change in the measured activities. By contrast, beta-apo-8'-carotenal increased the liver content of cytochrome P450, the activity of NADH- and NADPH-cytochrome c reductase, and strongly increased some cytochrome P450-dependent activities, particularly ethoxyresorufin O-deethylase (x158), methoxyresorufin O-demethylase (x22), pentoxy- and benzoxyresorufin O-dealkylases, but did not affect erythromycin N-demethylase nor nitrosodimethylamine N-demethylase activities. Phase II p-nitrophenol- and 4-hydroxy- biphenyl-uridine diphosphoglucuronosyl transferase activities were also increased by beta-apo-8'carotenal. Western blots of microsomal proteins clearly showed the induction of CYP1A1 and 1A2 by beta-apo-8'-carotenal. This induction profile resembles that produced by two other carotenoids: canthaxanthin and astaxanthin. Ethyl beta-apo-8'-carotenoate and citranaxanthin showed similar effects to beta-apo-8'-carotenal but of less intensity. 3. Three carotenoids: beta-apo-8'-carotenal, canthaxanthin and astaxanthin, are inducers of CYP1A1 and 1A2 in the rat. These carotenoids form a new class of inducers of CYP1A, structurally very different from the classical inducers as 3-methylcholanthrene, beta-naphtoflavone or dioxin.
Sigma-Aldrich
trans-β-Apo-8′-carotenal, ≥96.0% (UV)
Sigma-Aldrich
trans-β-Apo-8′-carotenal, ~20% apocarotenal basis (UV-vis), suspension (oily)