Merck
Ascorbic acid glycation: the reactions of L-threose in lens tissue.
Experimental eye research (1994-06-01)
B J Ortwerth, J A Speaker, M Prabhakaram, M G Lopez, E Y Li, M S Feather
ABSTRACT
L-Threose is a significant degradation product of ascorbic acid at pH 7.0 in the presence of oxygen. When compared to several other ascorbate-derived degradation products, it had the greatest ability to glycate and crosslink lens proteins in vitro. To determine whether L-threose was formed in the lens, the sugars in a TCA-soluble extract from human lenses were reduced to polyols with NaBH4, acetylated and analysed by gas-liquid chromatography. The threitol levels measured were 3.4 +/- 0.8 micrograms per lens (n = 4). GC-MS measurements made after reduction with NaBD4 indicated that threitol, but little or no threose, was originally present in the human lens. Rat lenses were incubated with [1-13C]D-threose for 24 hr, and considerable D-threitol formation was seen by NMR spectroscopy. Analysis of the lenses after medium removal showed that only [1-13C]threitol was present within the lenses indicating a rapid reduction of threose within the lens, presumably by aldose reductase. Assays with human recombinant aldose reductase and with human lens cortical and nuclear extracts all exhibited sorbinil-inhibitable aldose reductase activity with L-threose as substrate. This was confirmed by incubating a preparation of [1-14C]L-tetrose (a mixture of 40% L-threose and 45% L-erythrose) with both the pure aldose reductase and crude lens extracts followed by the subsequent identification of the [1-14C]L-threitol formed by thin layer chromatography. L-Threose degrades very slowly in 0.1 M phosphate buffer at pH 7.0, but the addition of a four-fold excess of N alpha-acetyl-L-lysine accelerated the rate of disappearance of threose 30-fold, indicating a rapid glycation reaction. When [1-14C]L-tetrose was incubated with a complete bovine lens homogenate, a linear incorporation into protein was observed over a 24 hr period. Increasing levels of lens extract exhibited increasing incorporation into protein. These data confirm a rapid reactivity of L-threose with lens protein and argue that glycation would occur in vivo in spite of the presence of aldose reductase.
Sigma-Aldrich
DL-Threitol, 97%
Sigma-Aldrich
D-Threitol, 99%