Merck
  • Enhancement of formation of the esophageal carcinogen benzylmethylnitrosamine from its precursors by Candida albicans.
Enhancement of formation of the esophageal carcinogen benzylmethylnitrosamine from its precursors by Candida albicans.
Proceedings of the National Academy of Sciences of the United States of America (1981-03-01)
C C Hsia, T T Sun, Y Y Wang, L M Anderson, D Armstrong, R A Good
ABSTRACT
Previous studies in Linxian, an area of China with a high incidence of esophageal carcinoma, showed that fungal infections are common in the esophageal epithelium of patients with either premalignant changes or early esophageal carcinoma. Fungi of the genus Candida were the most frequent invaders. In these areas nitrate and nitrite are often present in high concentrations in drinking water and staple grains. The present studies have established the ability of Candida albicans to augment the nitrosative formation of the esophagus-specific carcinogen, benzylmethylnitrosamine (NBMA; N-nitroso-N-methylbenzylamine). Stationary C. albicans cultures, with pH held at 6.8, were incubated with the precursors of NBMA, benzylmethylamine (BMA; N-methylbenzylamine) and NaNO(2). There was a significant increase in the amount of NBMA formed in these cultures, compared to precursors-only controls. The amount of NBMA synthesized depended on fungal cell number. Exponentially growing cultures were also able to cause NBMA formation. The identity of the NBMA was confirmed by high-performance liquid chromatographic coelution with authentic NBMA in three solvent systems and by mass spectroscopy. Boiled cells and conditioned medium in which cells had been incubated were not effective in enhancing nitrosation. Cultured Candida released acidic metabolites that reduced the pH of the medium when only a low concentration of buffer was present. Spontaneous nitrosation of BMA was enhanced under these acidic conditions. Thus, C. albicans infecting the esophageal epithelium could cause local formation of NBMA by both cell-mediated catalysis and extracellular decrease in pH.
Sigma-Aldrich
N-Benzylmethylamine, 97%
Pricing and availability is not currently available.