- Fluorometric measurement of glycosylated albumin in human serum.
Fluorometric measurement of glycosylated albumin in human serum.
Clinica chimica acta; international journal of clinical chemistry (1985-06-30)
Y Hayashi, M Makino
PMID4028431
ABSTRACT
A simple and rapid fluorometric assay of glycosylated albumin was developed using dansylated phenyl boronic acid (N-(5-dimethyl amino-1-naphthalene sulfonyl)-3-aminobenzene boronic acid). The emission spectrum of the reagent changes upon binding to cis-diols present on glycosylated albumin in serum, allowing quantitation of glycosylated albumin. Results from the new procedure correlated well with those from commercially prepared affinity chromatography (r = 0.85).
Sigma-Aldrich
3-(Dansylamino)phenylboronic acid, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)