Merck
  • The still unknown azobenzene - wavelength dependent photoanisotropy in amorphous azobenzene polymers.
The still unknown azobenzene - wavelength dependent photoanisotropy in amorphous azobenzene polymers.
Optics express (2013-02-08)
P S Ramanujam, Ole Bjarlin Jensen, Peter Tidemand-Lichtenberg
ABSTRACT
We demonstrate a new type of anisotropy in thin films of amorphous azobenzene polymers induced between 570 and 633 nm, where the absorbance in the film is on the order of 0.05. The anisotropy has a pronounced radial contribution. This observation points to an additional mechanism for the alignment of azobenzene molecules.
Sigma-Aldrich
Azobenzene, 98%
Pricing and availability is not currently available.