Merck
  • Invasive pulmonary aspergillosis due to Emericella nidulans var. echinulata, successfully cured by voriconazole and micafungin.
Invasive pulmonary aspergillosis due to Emericella nidulans var. echinulata, successfully cured by voriconazole and micafungin.
Journal of clinical microbiology (2013-02-01)
Jin Yu, Xiangdong Mu, Ruoyu Li
ABSTRACT
A 78-year-old male who was undergoing prolonged glucocorticoid treatment experienced cough and expectoration for 2 weeks. Galactomannan antigen analysis and a chest computed tomography (CT) scan suggested a diagnosis of invasive pulmonary aspergillosis. DNA sequencing indicated that Emericella nidulans var. echinulata was the causative agent. A combination of voriconazole and micafungin successfully treated the illness.
Sigma-Aldrich
D-Galacto-D-mannan from Ceratonia siliqua, ~95% (HPLC)