Merck
  • A kinetic study of the pH-dependent properties of the ferric undecapeptide of cytochrome c (microperoxidase).
A kinetic study of the pH-dependent properties of the ferric undecapeptide of cytochrome c (microperoxidase).
European journal of biochemistry (1977-07-01)
M T Wilson, R J Ranson, P Masiakowski, E Czarnecka, M Brunori
PMID20304
ABSTRACT
The ferric form of the haem undecapeptide, derived from horse cytochrome c by peptic digestion, undergoes at least three pH-induced transitions with pK values of 3.4, 5.8 and 7.6. Temperature-jump experiments suggest that the first of these is due to the binding of a deprotonated imidazole group to the feric iron while the second and third arise from the binding of the two available amino groups present (the alpha-NH2 of valine and the epsilon-NH2 of lysine). Molecular models indicate that steric retraints on the peptide dictate that these amino groups may only coordinate to iron atoms via intermolecular bonds, thus leading to the polymerization of the peptide. Cyanide binding studies are in agreement with these conclusions and also yield a value of 3.6 X 10(6) M-1 s-1 for the intrinsic combination constant of CN- anion with the haem. A model is proposed which describes the pH-dependent properties of the ferric undecapeptide.
Sigma-Aldrich
Microperoxidase disodium salt from horse heart, ≥85% (HPCE)