- Activity of plazomicin (ACHN-490) against MDR clinical isolates of Klebsiella pneumoniae, Escherichia coli, and Enterobacter spp. from Athens, Greece.
Activity of plazomicin (ACHN-490) against MDR clinical isolates of Klebsiella pneumoniae, Escherichia coli, and Enterobacter spp. from Athens, Greece.
Journal of chemotherapy (Florence, Italy) (2012-10-09)
Irene Galani, Maria Souli, George L Daikos, Zoi Chrysouli, Garyphalia Poulakou, Mina Psichogiou, Theofano Panagea, Athina Argyropoulou, Ioanna Stefanou, George Plakias, Helen Giamarellou, George Petrikkos
PMID23040681
ABSTRACT
The in vitro activity of plazomicin was evaluated against 300 multidrug resistant (MDR) (carbapenemase and/or ESBL-producing) isolates from four hospitals in Athens, an area where carbapenemase-producing organisms are endemic. Most of the isolates were also resistant to the legacy aminoglycosides with the MIC₅₀/MIC₉₀ to tobramycin, amikacin and gentamicin being 32/>32, 32/>32 and 4/>8 μg/ml, respectively. ACHN-490 retained activity (MICs ≤ 4 μg/ml) against all isolates of Klebsiella pneumoniae, Escherichia coli, and Enterobacter spp. tested with MIC₅₀ and MIC₉₀ of 1 and 2 μg/ml, respectively, irrespective of their MDR phenotype and it represents a promising alternative for the treatment of the most problematic Gram-negative pathogens.
MATERIALS