Supelco
3,5-Di-tert-butyl-4-hydroxytoluene, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, ≥97%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Butylhydroxytoluene, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
(−)-Linalool, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Benzyl alcohol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
cis-3,7-Dimethyl-2,6-octadien-1-ol, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
SKU
Pack Size
Availability
Price
Quantity
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzyl alcohol, puriss., meets analytical specification of Ph. Eur., BP, NF, 99-100.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Linalool, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Acetoin, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetoin, natural, ≥95%, FG
SKU
Pack Size
Availability
Price
Quantity
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Geraniol, ≥97%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetoin, primarily dimer, ≥95%, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Geraniol, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Linalool, 97%