Merck
  • Quercetin and sesamin protect neuronal PC12 cells from high-glucose-induced oxidation, nitrosative stress, and apoptosis.
Quercetin and sesamin protect neuronal PC12 cells from high-glucose-induced oxidation, nitrosative stress, and apoptosis.
Rejuvenation research (2012-04-25)
Julie Bournival, Marc-André Francoeur, Justine Renaud, Maria-Grazia Martinoli
ABSTRACT
Complications of diabetes are now well-known to affect sensory, motor, and autonomic nerves. Diabetes is also thought to be involved in neurodegenerative processes characteristic of several neurodegenerative diseases. Indeed, it has been acknowledged recently that hyperglycemia-induced oxidative stress contributes to numerous cellular reactions typical of central nervous system deterioration. The goal of the present study was to evaluate the effects of the polyphenol quercetin and the lignan sesamin on high-glucose (HG)-induced oxidative damage in an in vitro model of dopaminergic neurons, neuronal PC12 cells. When incubated with HG (13.5 mg/mL), neuronal PC12 cells showed a significant increase of cellular death. Our results revealed that quercetin and sesamin defend neuronal PC12 cells from HG-induced cellular demise. An elevated level of reactive oxygen and nitrogen species is a consequence of improved oxidative stress after HG administration, and we demonstrated that this production diminishes with quercetin and sesamin treatment. We also found that quercetin and sesamin elicited an increment of superoxide dismutase activity. DNA fragmentation, Bax/Bcl-2 ratio, nuclear translocation of apoptosis-inducing factor, as well as poly(adenosine diphosphate [ADP]-ribose) polymerase cleavage were significantly reduced by quercetin and sesamin administration, affirming their antiapoptotic features. Also, HG treatment impacted caspase-3 cleavage, supporting caspase-3-dependent pathways as mechanisms of apoptotic death. Our results indicate a powerful role for these natural dietary compounds and emphasize preventive or complementary nutritional strategies for diabetes control.
Product Description
Supelco
Sesamin, analytical standard
Pricing and availability is not currently available.