Merck
  • The endoplasmic reticulum is a target organelle for trivalent dimethylarsinic acid (DMAIII)-induced cytotoxicity.
The endoplasmic reticulum is a target organelle for trivalent dimethylarsinic acid (DMAIII)-induced cytotoxicity.
Toxicology and applied pharmacology (2012-03-20)
Hua Naranmandura, Shi Xu, Shota Koike, Li Qiang Pan, Bin Chen, Yan Wei Wang, Kanwal Rehman, Bin Wu, Zhe Chen, Noriyuki Suzuki
ABSTRACT
The purpose of present study was to characterize the endoplasmic reticulum stress and generation of ROS in rat liver RLC-16 cells by exposing to trivalent dimethylarsinous acid (DMAIII) and compared with that of trivalent arsenite (iAsIII) and monomethylarsonous acid (MMAIII). Protein kinase-like endoplasmic reticulum kinase (PERK) phosphorylation was significantly induced in cells exposed to DMAIII, while there was no change in phosphorylated PERK (P-PERK) detected in cells after exposure to iAsIII or MMAIII. The generation of reactive oxygen species (ROS) after DMAIII exposure was found to take place specifically in the endoplasmic reticulum (ER), while previous reports showed that ROS was generated in mitochondria following exposure to MMAIII. Meanwhile, cycloheximide (CHX) which is an inhibitor of protein biosynthesis strongly inhibited the DMAIII-induced intracellular ROS generation in the ER and the phosphorylation of PERK, suggesting the induction of ER stress probably occurs through the inhibition of the protein folding process. Activating transcription factor 4 (ATF4) and C/EBP homologous protein (CHOP) mRNA were induced by all three arsenic species, however, evidence suggested that they might be induced by different pathways in the case of iAsIII and MMAIII. In addition, ER resident molecular chaperone glucose-regulated protein78 (GRP78) was not affected by trivalent arsenicals, while it was induced in positive control only at high concentration (Thapsigargin;Tg), suggesting the GRP78 is less sensitive to low levels of ER stress. In summary, our findings demonstrate that the endoplasmic reticulum is a target organelle for DMAIII-induced cytotoxicity.
Sigma-Aldrich
Cacodylic acid, ≥99.0%
Sigma-Aldrich
Cacodylic acid, ≥98%
Cacodylic acid, analytical standard