Merck
  • Caco-2 cells cytotoxicity of nifuroxazide derivatives with potential activity against Methicillin-resistant Staphylococcus aureus (MRSA).
Caco-2 cells cytotoxicity of nifuroxazide derivatives with potential activity against Methicillin-resistant Staphylococcus aureus (MRSA).
Toxicology in vitro : an international journal published in association with BIBRA (2012-01-31)
Mariane B Fernandes, José E Gonçalves, Marcus T Scotti, Alex A de Oliveira, Leoberto C Tavares, Sílvia Storpirtis
ABSTRACT
It is important to determine the toxicity of compounds and co-solvents that are used in cell monolayer permeability studies to increase confidence in the results obtained from these in vitro experiments. This study was designed to evaluate the cytotoxicity of new nifuroxazide derivatives with potential activity against Methicillin-resistant Staphylococcus aureus (MRSA) in Caco-2 cells to select analogues for further in vitro permeability analyses. In this study, nitrofurantoin and nifuroxazide, in addition to 6 furanic and 6 thiophenic nifuroxazide derivatives were tested at 2, 4, 6, 8 and 10 μg/mL. In vitro cytotoxicity assays were performed according to the MTT (methyl tetrazolium) assay protocol described in ISO 10993-5. The viability of treated Caco-2 cells was greater than 83% for all tested nitrofurantoin concentrations, while those treated with nifuroxazide at 2, 4 and 6 μg/mL had viabilities greater than 70%. Treatment with the nifuroxazide analogues resulted in viability values greater than 70% at 2 and 4 μg/mL with the exception of the thiophenic methyl-substituted derivative, which resulted in cell viabilities below 70% at all tested concentrations. Caco-2 cells demonstrated reasonable viability for all nifuroxazide derivatives, except the thiophenic methyl-substituted compound. The former were selected for further permeability studies using Caco-2 cells.