Merck
  • In vitro and in vivo efficacy of Monepantel (AAD 1566) against laboratory models of human intestinal nematode infections.
In vitro and in vivo efficacy of Monepantel (AAD 1566) against laboratory models of human intestinal nematode infections.
PLoS neglected tropical diseases (2012-01-05)
Lucienne Tritten, Angelika Silbereisen, Jennifer Keiser
ABSTRACT
Few effective drugs are available for soil-transmitted helminthiases and drug resistance is of concern. In the present work, we tested the efficacy of the veterinary drug monepantel, a potential drug development candidate compared to standard drugs in vitro and in parasite-rodent models of relevance to human soil-transmitted helminthiases. A motility assay was used to assess the efficacy of monepantel, albendazole, levamisole, and pyrantel pamoate in vitro on third-stage larvae (L3) and adult worms of Ancylostoma ceylanicum, Necator americanus and Trichuris muris. Ancylostoma ceylanicum- or N. americanus-infected hamsters, T. muris- or Ascaris suum-infected mice, and Strongyloides ratti-infected rats were treated with single oral doses of monepantel or with one of the reference drugs. Monepantel showed excellent activity on A. ceylanicum adults (IC(50) = 1.7 µg/ml), a moderate effect on T. muris L3 (IC(50) = 78.7 µg/ml), whereas no effect was observed on A. ceylanicum L3, T. muris adults, and both stages of N. americanus. Of the standard drugs, levamisole showed the highest potency in vitro (IC(50) = 1.6 and 33.1 µg/ml on A. ceylanicum and T. muris L3, respectively). Complete elimination of worms was observed with monepantel (10 mg/kg) and albendazole (2.5 mg/kg) in A. ceylanicum-infected hamsters. In the N. americanus hamster model single 10 mg/kg oral doses of monepantel and albendazole resulted in worm burden reductions of 58.3% and 100%, respectively. Trichuris muris, S. ratti and A. suum were not affected by treatment with monepantel in vivo (following doses of 600 mg/kg, 32 mg/kg and 600 mg/kg, respectively). In contrast, worm burden reductions of 95.9% and 76.6% were observed following treatment of T. muris- and A. suum infected mice with levamisole (200 mg/kg) and albendazole (600 mg/kg), respectively. Monepantel reveals low or no activities against N. americanus, T. muris, S. ratti and A. suum in vivo, hence does not qualify as drug development candidate for human soil-transmitted helminthiases.
Sigma-Aldrich
Aminoacetonitrile, ≥98%