Merck
  • Acridine yellow G blocks glioblastoma growth via dual inhibition of epidermal growth factor receptor and protein kinase C kinases.
Acridine yellow G blocks glioblastoma growth via dual inhibition of epidermal growth factor receptor and protein kinase C kinases.
The Journal of biological chemistry (2012-01-05)
Qi Qi, Kunyan He, Min-Heui Yoo, Chi-Bun Chan, Xia Liu, Zhaobin Zhang, Jeffrey J Olson, Ge Xiao, Liya Wang, Hui Mao, Haian Fu, Hui Tao, Suresh S Ramalingam, Shi-Yong Sun, Paul S Mischel, Keqiang Ye
ABSTRACT
Amplification of the epidermal growth factor receptor (EGFR), frequently expressed as a constitutively active deletion mutant (EGFRvIII), occurs commonly in glioblastoma multiformes (GBM). However, blockade of EGFR is therapeutically disappointing for gliomas with PTEN deletion. To search for small molecules treating this aggressive cancer, we have established a cell-based screening and successfully identified acridine yellow G that preferentially blocks cell proliferation of the most malignant U87MG/EGFRvIII cells over the less malignant U87MG/PTEN cells. Oral administration of this compound markedly diminishes the brain tumor volumes in both subcutaneous and intracranial models. It directly inhibits EGFR and PKCs with IC(50) values of ~7.5 and 5 μM, respectively. It dually inhibits EGFR and PKCs, resulting in a blockade of mammalian target of rapamycin signaling and cell cycle arrest in the G(1) phase, which leads to activation of apoptosis in the tumors. Hence, combinatorial inhibition of EGFR and PKCs might provide proof of concept in developing therapeutic agents for treating malignant glioma and other human cancers.
Sigma-Aldrich
Acridine Yellow G, Dye content 90 %
Pricing and availability is not currently available.