Merck
  • Increasing the thermal storage capacity of a phase change material by encapsulation: preparation and application in natural rubber.
Increasing the thermal storage capacity of a phase change material by encapsulation: preparation and application in natural rubber.
ACS applied materials & interfaces (2011-09-03)
Songpon Phadungphatthanakoon, Sirilux Poompradub, Supason P Wanichwecharungruang
ABSTRACT
Existing encapsulated organic phase change materials (PCM) usually contain a shell material that possesses a poor heat storage capacity and so results in a lowered latent heat storage density of the encapsulated PCM compared to unencapsulated PCM. Here, we demonstrate the use of a novel microencapsulation process to encapsulate n-eicosane (C20) into a 2:1 (w/w) ratio blend of ethyl cellulose (EC):methyl cellulose (MC) to give C20-loaded EC/MC microspheres with an increased heat storage capacity compared to the unencapsulated C20. Up to a 29 and 24% increase in the absolute enthalpy value during crystallization and melting were observed for the encap-C20/EC/MC microparticles with a 9% (w/w) EC/MC polymer content. The mechanism that leads to the increased latent heat storage capacity is discussed. The blending of the water-dispersible C20-loaded EC/MC microspheres into natural rubber latex showed excellent compatibility, and the obtained rubber composite showed not only an obvious thermoregulation property but also an improved mechanical property.
Product Description
Supelco
Eicosane, analytical standard
Sigma-Aldrich
Eicosane, 99%