Merck
  • Synthesis of apiose-containing oligosaccharide fragments of the plant cell wall: fragments of rhamnogalacturonan-II side chains A and B, and apiogalacturonan.
Synthesis of apiose-containing oligosaccharide fragments of the plant cell wall: fragments of rhamnogalacturonan-II side chains A and B, and apiogalacturonan.
Organic & biomolecular chemistry (2011-08-19)
Sergey A Nepogodiev, Margherita Fais, David L Hughes, Robert A Field
ABSTRACT
Fragments of pectic polysaccharides rhamnogalacturonan-II (RG-II) and apiogalacturonan were synthesised using p-tolylthio apiofuranoside derivatives as key building blocks. Apiofuranose thioglycosides can be conveniently prepared by cyclization of the corresponding dithioacetals possessing a 2,3-O-isopropylidene group, which is required for preservation of the correct (3R) configuration of the apiofuranose ring. The remarkable stability of this protecting group in apiofuranose derivatives requires its replacement with a more reactive protecting group, such as a benzylidene acetal which was used in the synthesis of trisaccharide β-Rhap-(1→3')-β-Apif-(1→2)-α-GalAp-OMe. The X-ray crystal structure of the protected precursor of this trisaccharide has been elucidated.
Sigma-Aldrich
D-Apiose solution, ≥99.0% (HPLC)