Merck
  • Synthesis of 1-aminoisoquinolines via Rh(III)-catalyzed oxidative coupling.
Synthesis of 1-aminoisoquinolines via Rh(III)-catalyzed oxidative coupling.
Organic letters (2011-08-05)
Xiaohong Wei, Miao Zhao, Zhengyin Du, Xingwei Li
ABSTRACT
[RhCp*Cl(2)](2) can catalyze the oxidative coupling of N-aryl and N-alkyl benzamidines with alkynes to give N-substituted 1-aminoisoquinolines in high selectivity.
Sigma-Aldrich
1-Aminoisoquinoline, 99%