Merck
  • Incorporation of nisin in poly (ethylene-co-vinyl acetate) films by melt processing: a study on the antimicrobial properties.
Incorporation of nisin in poly (ethylene-co-vinyl acetate) films by melt processing: a study on the antimicrobial properties.
Journal of food protection (2011-07-12)
Roberto Scaffaro, Luigi Botta, Sandra Marineo, Anna Maria Puglia
ABSTRACT
Both industry and academia have shown a growing interest in materials with antimicrobial properties suitable for food packaging applications. In this study, we prepared and characterized thin films of ethylene-co-vinyl acetate (EVA) copolymer with antimicrobial properties. The films were prepared with a film blowing process by incorporating a nisin preparation as an antimicrobial agent in the melt. Two grades of EVA containing 14 and 28% (wt/wt) vinyl acetate (EVA 14 and EVA 28, respectively) and two commercial formulations of nisin with different nominal activities were used. The effect of the nisin concentration also was evaluated. The films with the highest antimicrobial activity were those formulated with nisin at the highest activity and EVA with the highest content of vinyl acetate. The use of the commercial formulation of nisin with high activity in the EVA films allowed reduction in the amount of nisin needed to provide antimicrobial properties. Consequently, the mechanical properties of these films were only slightly inferior to those of the pure polymers. In contrast, films prepared by incorporating more of the nisin with lower activity had poor mechanical properties. The effect of different processing temperatures used in the preparation of the films on the antimicrobial properties of the films also was evaluated. The materials displayed antimicrobial properties even when they were prepared at temperatures as high as 160 °C, probably because of the very short processing time (60 to 90 s) required for preparation.
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 40 wt. %, melt index (41-63 dg/min (190°C/2.16kg)), contains 190-910 ppm inhibitor
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 18 wt. %, melt index 8 g/10 min (190°C/2.16kg), contains 200-900 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 12 wt. %, melt index 8 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Poly(ethylene-co-vinyl acetate), vinyl acetate 25 wt. %, melt index 19 g/10 min (190°C/2.16 kg), contains 200-900 ppm BHT as inhibitor