Merck
  • Molecular characterization and expression analysis of dihydroflavonol 4-reductase (DFR) gene in Saussurea medusa.
Molecular characterization and expression analysis of dihydroflavonol 4-reductase (DFR) gene in Saussurea medusa.
Molecular biology reports (2011-06-28)
Houhua Li, Jian Qiu, Fudong Chen, Xiaofen Lv, Chunxiang Fu, Dexiu Zhao, Xuejun Hua, Qiao Zhao
ABSTRACT
Dihydroflavonol 4-reductase (DFR), which catalyzes the reduction of dihydroflavonols to leucoanthocyanins, is a key enzyme in the biosynthesis of anthocyanidins, proanthocyanidins, and other flavonoids of importance in plant development and human nutrition. This study isolated a full length cDNA encoding DFR, designated as SmDFR (GenBank Accession No. EF600682), by screening a cDNA library from a red callus line of Saussurea medusa, which is an endangered, traditional Chinese medicinal plant with high pharmacological value. SmDFR was functionally expressed in yeast (Saccharomyces cerevisiae) to confirm that SmDFR can readily reduce dihydroquercetin (DHQ) and dihydrokampferol (DHK), but it could not reduce dihydromyricetin (DHM). The deduced SmDFR structure shared extensive sequence similarity with previously characterized plant DFRs and phylogenetic analysis showed that it belonged to the plant DFR super-family. SmDFR also possessed flavanone 4-reductase (FNR) activity and can catalyze the conversion of eridictyol to luteoforol. Real-time PCR analysis showed that the expression level of SmDFR was higher in flowers compared with both leaves and roots. This work greatly enhances our knowledge of flavonoid biosynthesis in S. medusa and marks a major advance that could facilitate future genetic modification of S. medusa.
Product Description
Supelco
Dihydrokaempferol, analytical standard
Pricing and availability is not currently available.