Merck
  • Interaction of calf thymus dsDNA with anti-tumor drug tamoxifen studied by zero current potentiometry.
Interaction of calf thymus dsDNA with anti-tumor drug tamoxifen studied by zero current potentiometry.
Biosensors & bioelectronics (2011-04-29)
Xiao-xia Guo, Zhang-jun Song, Jie-juan Sun, Jun-feng Song
ABSTRACT
Since the electrochemical oxidation peaks of both DNA and anti-tumor drug tamoxifen (TAM) overlapped with each other, the known electrochemical methods were limited in the study of the interactions between DNA and TAM. In this paper, zero current potentiometry, a new electrochemical method, was used to study the interaction of calf thymus dsDNA with TAM. The dsDNA was immobilized on the surface of carbon paste (dsDNA/CP). The dsDNA/CP connected in series between the clips of working and counter electrodes of a potentiostat and a reference electrode were immersed in aqueous solution containing TAM, the interaction of dsDNA with TAM produced a change in interfacial potential at the dsDNA/CP/solution interface. When linear sweep potential was applied to the dsDNA/CP and the corresponding I-E curve was recorded, interfacial potential offset applied potential partially, making the I-E curve displace along potential axis. Zero current potential where circuit current I was equal to zero in the I-E curve was measured to check the displacement of the I-E curve. Based on the displacement, the thermodynamic constants of the interaction between dsDNA and TAM were determined. The binding ratio of dsDNA with TAM was found to be 1:1 and the apparent binding constant was (6.85±0.20)×10(6) M(-1). As zero current potentiometry was independent of the changes in redox potential or current of both dsDNA and TAM themselves, the interaction was studied in their natural forms without damage. Moreover, TAM can be determined. The detection limit was 1.1×10(-7) M.
Sigma-Aldrich
Mercury(I) chloride, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Mercury(I) chloride, ACS reagent, ≥99.5%