Merck
  • Combined effect of N-acetyl cysteine, zinc, and selenium against chronic dimethylmercury-induced oxidative stress: a biochemical and histopathological approach.
Combined effect of N-acetyl cysteine, zinc, and selenium against chronic dimethylmercury-induced oxidative stress: a biochemical and histopathological approach.
Archives of environmental contamination and toxicology (2011-03-23)
Deepmala Joshi, Deepak Mittal, Sadhana Shrivastav, Sangeeta Shukla, Ajai Kumar Srivastav
ABSTRACT
Mercury (Hg), widely used in industry, is a great environmental health problem for humans and animals. Despite several reports regarding Hg toxicity, there is scarcity of data on its toxic manifestations on Sprague Dawley rats under realistic exposure conditions. Experimental studies have shown that sulphur-containing antioxidants have beneficial effects against the detrimental properties of Hg. The present work was aimed to study the therapeutic potential of combined administration of N-acetyl cysteine (NAC; 2 mmol/kg ip), zinc (Zn; 2 mmol/kg po), and selenium (Se; 0.5 mg/kg po) against dimethylmercury (DMM; 1 mg/kg po)-intoxicated male rats for 12 weeks. Exposure to DMM caused significant alterations in cytochrome P450 (CYP) activity, microsomal lipid peroxidation, and proteins. Activities of transaminases (aspartate aminotransferase/alanine aminotransferase), alkaline phosphatase, and lactate dehydrogenase in serum, as well as activities of CYP enzymes aniline hydroxylase (AH), amidopyrine-N-demethylase (AND) in liver microsomes and activities of acid phosphatase, alkaline phosphatase, glucose-6-phophatase, and succinic dehydrogenase in the liver and kidney, were significantly altered after DMM administration. DMM exposure also induced severe hepato-renal alterations at the histopathological level. NAC, along with Zn and Se, dramatically reversed the alterations in all of the variables more toward control. The study results conclude that protective intervention of combined treatment of NAC, along with Zn and Se, is beneficial in attenuating DMM-induced systemic toxicity.
Sigma-Aldrich
Dimethylmercury, 95%