Merck
  • Ovarian hormone deficiency reduces intrinsic excitability and abolishes acute estrogen sensitivity in hippocampal CA1 pyramidal neurons.
Ovarian hormone deficiency reduces intrinsic excitability and abolishes acute estrogen sensitivity in hippocampal CA1 pyramidal neurons.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2011-02-18)
Wendy W Wu, John P Adelman, James Maylie
ABSTRACT
Premature and uncompensated loss of ovarian hormones following ovariectomy (OVX) elevates the risks of cognitive impairment and dementia. These risks are prevented with estrogen (E(2))-containing hormone replacement therapy initiated shortly following OVX but not after substantial delay. Currently, the cellular bases underlying these clinical findings are unknown. At the cellular level, intrinsic membrane properties regulate the efficiency of synaptic inputs to initiate output action potentials (APs), thereby affecting neuronal communication, hence cognitive processing. This study tested the hypothesis that in CA1 pyramidal neurons, intrinsic membrane properties and their acute regulation by E(2) require ovarian hormones for maintenance. Whole-cell current-clamp recordings were performed on neurons from ∼ 7-month-old OVX rats that experienced either short-term (10 d, control OVX) or long-term (5 months, OVX(LT)) ovarian hormone deficiency. The results reveal that long-term hormone deficiency reduced intrinsic membrane excitability (IE) as measured by the number of evoked APs and firing duration for a given current injection. This was accompanied by AP broadening, an increased slow afterhyperpolarization (sAHP), and faster accumulation of Na(V) channel inactivation during repetitive firing. In the control OVX neurons, E(2) acutely increased IE and reduced the sAHP. In contrast, acute regulation of IE by E(2) was absent in the OVX(LT) neurons. Since the degree of IE of hippocampal pyramidal neurons is positively related with hippocampus-dependent learning ability, and modulation of IE is observed following successful learning, these findings provide a framework for understanding hormone deficiency-related cognitive impairment and the critical window for therapy initiation.
Sigma-Aldrich
CGP 55845 hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.