Merck
  • Novel chelating resin with cyanoguanidine group: useful recyclable materials for Hg(II) removal in aqueous environment.
Novel chelating resin with cyanoguanidine group: useful recyclable materials for Hg(II) removal in aqueous environment.
Journal of hazardous materials (2010-11-13)
Xiaojie Ma, Yanfeng Li, Zhengfang Ye, Liuqing Yang, Lincheng Zhou, Liyuan Wang
ABSTRACT
A novel chelating resin containing cyanoguanidine moiety has been successfully prepared by the functionalizing reaction of a macroporous bead based on chloromethylated copolymer of styrene-divinylbenzene (CMPS) with dicyandiamide (DCDA) in the presence of phase transfer catalyst. The Fourier transform-infrared spectra (FT-IR) and scanning electron microscopy (SEM) were employed in the characterization of the resulting chelating resin, meanwhile, the adsorption properties of the resin for Hg(II) were investigated by batch and column methods. The results indicated that the resin displayed a marked advantage in Hg(II) binding capacity, and the saturated adsorption capacity estimated from the Langmuir model was dramatically up to 1077 mg g(-1) at 45 °C. Furthermore, it was found that the resin was able to selectively separate Hg(II) from multicomponent solutions with Zn(II), Cu(II), Pb(II) and Mg(II). The desorption process of Hg(II) was tested with different eluents and the ratio of the highest recovery reached to 96% under eluting condition of 1M HCl+10% thiourea. Consequently, the resulting chelating resin would provide a potential application for treatment process of Hg(II) containing wastewater.
Sigma-Aldrich
Dicyandiamide, 99%
Supelco
Cyanoguanidine, pharmaceutical impurity standard, ≥95.0% (HPLC)